Learn More
BINDER™ Reinforced Rack
Product Code. 30238381 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB/KB PRO 1060, KBF/KBF PRO 1060
KB/KB PRO 720, KBF/KBF PRO 720
KB/KBF/KB PRO/KBF PRO 260/470
KBF 130, KBF PRO 130
Unit Size:
Each
Product Code. 30238381
Brand: BINDER™ 80122414
Only null left
Add to basket
Add to basket
Reinforced shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
  • Shelf rack made of stainless steel
  • Includes set of U-rails
Specifications
For Use With (Equipment) KB/KB PRO 720, KBF/KBF PRO 720
Material Stainless Steel
Type Rack