Learn More
Gilac™ Reinforced Large Volume Container
Description
- Excellent mechanical resistance: made from high density polyethylene for increased shock resistance.
- The durable, thick walls mean no distortion and warping.
- Limited gaps and overlapping, equipped with edges so easy to handle.
- Volume in litres is equivalent to the container filled to the brim.
Specifications
Specifications
| Type | Storage Tote |
| Material | HDPE |
| Color | White |
| Shape | Rectangular |
| Capacity (English) | 58.1 gal. |
| Capacity (Metric) | 220 L |
| Height (English) | 2.4 ft. |
| Height (Metric) | 720 mm |