Learn More
Buerkle™ Reduced Nipple with Outer Thread
Manufactured from polypropylene.
6.15 EUR - 7.80 EUR
Specifications
Product Type | Nipple Reduced Outer |
---|---|
Certifications/Compliance | EU Foodstuffs |
Autoclavable | Autoclavable |
Material | Polypropylene |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
10240562
|
Buerkle™
8506-1234 |
13.5 mm |
6.15 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10054940
|
Buerkle™
8506-3410 |
18 mm |
7.80 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
Nipple Reduced Outer | |
EU Foodstuffs | |
Autoclavable | |
Polypropylene |