Learn More
Buerkle™ Reduced Nipple with Outer Thread
Manufactured from polypropylene.
Brand:  Buerkle™ 8506-3410
7.29 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.01250kg
 View more versions of this product
Product Code. 10054940
  • 7.80 EUR / Each
Estimated Shipment: 29-07-2024
Description
  • Polypropylene construction
  • Autoclavable to 121°C
  • Safe for use with foodstuffs
  • Supplied in packs of 10
Specifications
Specifications
Nipple Reduced Outer
Autoclavable
3/4 to 1 in.
EU Foodstuffs
Polypropylene
18 mm