Learn More
Tenak™ Red Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
93.15 EUR - 172.00 EUR
Specifications
Color Red
Material Cardboard
Shape Square
Type Cryo Box
View More Specs
Products 6
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
15192105
View Documents Promotion Details
Tenak™
TE22542
136 x 136 x 100 mm
167.00 EUR
Pack of 36
View alternative products
Save Up to 
»
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
11591655
View Documents Promotion Details
Tenak™
TE22084
133 x 133 x 130 mm
167.00 EUR
Pack of 36
View alternative products
Save Up to 
»
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
15210614
View Documents Promotion Details
Tenak™
TE22562
136 x 136 x 130 mm
172.00 EUR
Pack of 36
View alternative products
Save Up to 
»
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
15200524
View Documents Promotion Details
Tenak™
TE22002
133 x 133 x 32 mm
93.15 EUR
Pack of 36
View alternative products
Save Up to 
»
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
15270594
missing translation for 'viewDocuments' Promotion Details
Tenak™
TE22502
136 x 136 x 50 mm
95.40 EUR
Pack of 36
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 19-07-2024
to see stock.
Zum Warenkorb hinzufügen
15240524
missing translation for 'viewDocuments' Promotion Details
Tenak™
TE22060
133 x 133 x 75 mm
110.00 EUR
Pack of 36
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 19-07-2024
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Spezifikation
Spezifikation
Red
Cardboard
Square
Cryo Box
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate