Learn More
Tenak™ Red Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Brand:  Tenak™ TE22060
Additional Details : Weight : 3.20000kg
 View more versions of this product
Product Code. 15240524
  • 110.00 EUR / Pack of 36
Estimated Shipment: 05-08-2024
Description
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Specifications
Specifications
Red
Square
133 x 133 x 75 mm
Cardboard
Cryo Box