missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Recombinant Streptavidin

Product Code. 17126781
Click to view available options
:
5 mg

Product Code. 17126781

Brand: ACROBiosystems STNN51165mg

Please to purchase this item. Need a web account? Register with us today!

Recombinant Protein

Streptavidin is a tetrameric protein purified from the bacterium Streptomyces avidinii, and exhibits high binding affinity for biotin. Able to bind one molecule of biotin with each subunit. Streptavidin (PI=6.0-7.5) has lower level of non-specific binding to various biological components at physiological pH than avidin (PI=7.4), resulting from its isoelectric point (PI).Streptavidin is useful in affinity chromatography, ELISA, immunohistochemistry and Western Blotting.

This protein .

Specifications

Quantity 5 mg
Source Recombinant Streptavidin (STN-N5116) is expressed from E.coli cells.
Storage Requirements -20°C
Gene Alias Streptavidin,SA
Common Name Streptavidin
Conjugate Unconjugated
Buffer NaCl, pH7.4
Expression System E.coli
Form Powder
Protein Subtype Other Recombinant Proteins
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.