missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IFNG / Interferon Gamma Protein
A DNA sequence encoding the Ovis aries (Sheep) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6206-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001009803.1 | |
IFNG / Interferon Gamma Protein | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
Yeast | |
QGPFFKEIEN LKEYFNASNP DVAKGGPLFS EILKNWKEES DKKIIQSQIV SFYFKLFENL KDNQVIQRSM DIIKQDMFQK FLNGSSEKLE DFKRLIQIPV DDLQIQRKAI NELIKVMNDL SPKSNLRKRK RSQNLFRGRR ASM | |
Greater than 90% as determined by SDS-PAGE. |
443396 | |
1 mg | |
Not determined. | |
Full Length | |
Sheep | |
His | |
Tris-based buffer, 50% glycerol |