missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IFNG / Interferon Gamma Protein
A DNA sequence encoding the Ovis aries (Sheep) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.
352.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_001009803.1 |
---|---|
Gene ID (Entrez) | 443396 |
Name | IFNG / Interferon Gamma Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031560
|
enQuireBio™
QP6206-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071560
|
enQuireBio™
QP6206-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041560
|
enQuireBio™
QP6206-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061560
|
enQuireBio™
QP6206-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081560
|
enQuireBio™
QP6206-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051560
|
enQuireBio™
QP6206-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001009803.1 | |
IFNG / Interferon Gamma Protein | |
Not determined. | |
Full Length | |
Sheep | |
His | |
Tris-based buffer, 50% glycerol |
443396 | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
Yeast | |
QGPFFKEIEN LKEYFNASNP DVAKGGPLFS EILKNWKEES DKKIIQSQIV SFYFKLFENL KDNQVIQRSM DIIKQDMFQK FLNGSSEKLE DFKRLIQIPV DDLQIQRKAI NELIKVMNDL SPKSNLRKRK RSQNLFRGRR ASM | |
Greater than 90% as determined by SDS-PAGE. |