Learn More
R&D Systems™ Recombinant SARS-CoV-2 Spike S1 Subunit Biotin Protein
Product Code. 30000916
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. 30000916
Brand: R&D Systems™ BT10569050
His-tag. Biotinylated via Amines
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN).
TRUSTED_SUSTAINABILITY
Specifications
Accession Number YP_009724390.1
Conjugate Biotin
For Use With (Application) Bioactivity
Quantity 50 μg
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Synonym SARS-CoV-2
Target SARS-CoV-2
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.