missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PLA2G2A Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PLA2G2A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7058-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_113786.3 | |
PLA2G2A Protein | |
Research Use Only | |
Pla2g2a | |
Recombinant Protein | |
E. coli | |
SLLEFGQMIL FKTGKRADVS YGFYGCHCGV GGRGSPKDAT DWCCVTHDCC YNRLEKRGCG TKFLTYKFSY RGGQISCSTN QDSCRKQLCQ CDKAAAECFA RNKKSYSLKY QFYPNKFCKG KTPSC | |
Greater than 90% as determined by SDS-PAGE. |
29692 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |