missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PLA2G2A Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PLA2G2A, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_113786.3 |
---|---|
Gene ID (Entrez) | 29692 |
Name | PLA2G2A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077250
|
enQuireBio™
QP7058-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097250
|
enQuireBio™
QP7058-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087250
|
enQuireBio™
QP7058-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007260
|
enQuireBio™
QP7058-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017260
|
enQuireBio™
QP7058-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027260
|
enQuireBio™
QP7058-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_113786.3 | |
PLA2G2A Protein | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
29692 | |
Research Use Only | |
Pla2g2a | |
Recombinant Protein | |
E. coli | |
SLLEFGQMIL FKTGKRADVS YGFYGCHCGV GGRGSPKDAT DWCCVTHDCC YNRLEKRGCG TKFLTYKFSY RGGQISCSTN QDSCRKQLCQ CDKAAAECFA RNKKSYSLKY QFYPNKFCKG KTPSC | |
Greater than 90% as determined by SDS-PAGE. |