missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat KNG1 / BDK / kininogen-1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) KNG1 / BDK / kininogen-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9238-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036873.1 | |
KNG1 / BDK / kininogen-1 Protein | |
Research Use Only | |
Kng1 | |
Recombinant Protein | |
Yeast | |
APRVKKPKES TTVSPSYIAR VQEERDPGNE QGPIHGHGWL HAKQIKNKNH QGHKHGHGIG HGHQKPHGLG HGHQLKLDDL KQQREDGYDH RHPVGHGHGQ RHGHGHGHGH GRDKHTNKDK NNVKHTDQRR APLTSSSEDN TTSTQIQGRT EGFTLNPPLA QPAVISRGFQ DSGFTEGVIA TTSPYDTETH DDLIPDIHVQ PDSLSFKLIS DFPEATSHKC PGRPWKPVSR KDPTIETTEF SDFDLLDALS | |
Greater than 90% as determined by SDS-PAGE. |
25087 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |