missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat KNG1 / BDK / kininogen-1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) KNG1 / BDK / kininogen-1, was expressed in the hosts and tags indicated.
245.00 EUR - 2645.00 EUR
Specifications
Accession Number | NP_036873.1 |
---|---|
Gene ID (Entrez) | 25087 |
Name | KNG1 / BDK / kininogen-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16067891
|
enQuireBio™
QP9238-YE-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007901
|
enQuireBio™
QP9238-YE-500UG |
500 μg |
1925.00 EUR
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077891
|
enQuireBio™
QP9238-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097891
|
enQuireBio™
QP9238-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017901
|
enQuireBio™
QP9238-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087891
|
enQuireBio™
QP9238-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_036873.1 | |
KNG1 / BDK / kininogen-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
25087 | |
Research Use Only | |
Kng1 | |
Recombinant Protein | |
Yeast | |
APRVKKPKES TTVSPSYIAR VQEERDPGNE QGPIHGHGWL HAKQIKNKNH QGHKHGHGIG HGHQKPHGLG HGHQLKLDDL KQQREDGYDH RHPVGHGHGQ RHGHGHGHGH GRDKHTNKDK NNVKHTDQRR APLTSSSEDN TTSTQIQGRT EGFTLNPPLA QPAVISRGFQ DSGFTEGVIA TTSPYDTETH DDLIPDIHVQ PDSLSFKLIS DFPEATSHKC PGRPWKPVSR KDPTIETTEF SDFDLLDALS | |
Greater than 90% as determined by SDS-PAGE. |