missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat GPT Protein

Product Code. 15994528
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15994528

Brand: enQuireBio™ QP106535ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GPT was constructed and used to recombinantly synthesize the protein.

Specifications

Name GPT Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Specific activity is > 60units/mg, and is defined as the amount of enzyme that cleaves 1umole of L-Alanine to L-Glutamate per minute at pH 7.5 at 37C.
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMASRVND QSQASRNGLK GKVLTLDTMN PCVRRVEYAV RGPIVQRALE LEQELRQGVK KPFTEVIRAN IGDAQAMGQR PITFFRQVLA LCVYPNLLSS PDFPEDAKRR AERILQACGG HSLGAYSISS GIQPIREDVA QYIERRDGGI PADPNNIFLS TGASDAIVTM LKLLVSGEGR ARTGVLIPIP QYPLYSAALA ELDAVQVDYY LDEERAWALD IAELRRALCQ ARDRCCPRVL CVINPGNPTG QVQTRECIEA VIRFAFKEGL FLMADEVYQD NVYAEGSQFH SFKKVLMEMG PPYSTQQELA SFHSVSKGYM GECGFRGGYV EVVNMDAEVQ KQMGKLMSVR LCPPVPGQAL MDMVVSPPTP SEPSFKQFQA ERQEVLAELA AKAKLTEQVF NEAPGIRCNP VQGAMYSFPQ VQLPLKAVQR AQELGLAPDM FFCLCLLEET GICVVPGSGF GQQEGTYHFR MTILPPMEKL RLLLEKLSHF HAKFTHEYS
Buffer GPT protein solution (1 mg/ml) containing Phosphate Buffered Saline pH 7.4 and 10% glycerol.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.