missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pseudomonas Creatinine amidohydrolase Protein

A DNA sequence encoding the Pseudomonas putida (Arthrobacter siderocapsulatus) Creatinine amidohydrolase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6914-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006730

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Pseudomonas Creatinine amidohydrolase Protein
Research Use Only
crnA
Recombinant Protein
E. coli
MSKSVFVGEL TWKEYEARVA AGDCVLMLPV GALEQHGHHM CMNVDVLLPT AVCKRVAERI GALVMPGLQY GYKSQQKSGG GNHFPGTTSL DGATLTGTVQ DIIRELARHG ARRLVLMNGH YENSMFIVEG IDLALRELRY AGIQDFKVVV LSYWDFVKDP AVIQQLYPEG FLGWDIEHGG VFETSLMLAL YPDLVDLDRV VDHPPATFPP YDVFPVDPAR TPAPGTLSSA KTASREKGEL ILEVCVQGIA DAIREEFPPT
Greater than 90% as determined by SDS-PAGE.
50 μg
Not determined.
Full Length
seudomonas
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers