missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pseudomonas Creatinine amidohydrolase Protein
A DNA sequence encoding the Pseudomonas putida (Arthrobacter siderocapsulatus) Creatinine amidohydrolase, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Name | Pseudomonas Creatinine amidohydrolase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | crnA |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16056720
|
enQuireBio™
QP6914-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076720
|
enQuireBio™
QP6914-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066720
|
enQuireBio™
QP6914-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086720
|
enQuireBio™
QP6914-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096720
|
enQuireBio™
QP6914-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006730
|
enQuireBio™
QP6914-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Pseudomonas Creatinine amidohydrolase Protein | |
Not determined. | |
Full Length | |
seudomonas | |
GST | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
crnA | |
Recombinant Protein | |
E. coli | |
MSKSVFVGEL TWKEYEARVA AGDCVLMLPV GALEQHGHHM CMNVDVLLPT AVCKRVAERI GALVMPGLQY GYKSQQKSGG GNHFPGTTSL DGATLTGTVQ DIIRELARHG ARRLVLMNGH YENSMFIVEG IDLALRELRY AGIQDFKVVV LSYWDFVKDP AVIQQLYPEG FLGWDIEHGG VFETSLMLAL YPDLVDLDRV VDHPPATFPP YDVFPVDPAR TPAPGTLSSA KTASREKGEL ILEVCVQGIA DAIREEFPPT | |
Greater than 90% as determined by SDS-PAGE. |