missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig Antibacterial peptide PMAP-36 Protein

A DNA sequence encoding the Sus scrofa (Pig) Antibacterial peptide PMAP-36, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7219-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037770

  • 2985.00 EUR / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001123437.1
Antibacterial peptide PMAP-36 Protein
Research Use Only
PMAP36
Recombinant Protein
E. coli
VGRFRRLRKK TRKRLKKIGK VLKWIPPIVG SIPLGCG
Greater than 90% as determined by SDS-PAGE.
100170124
1 mg
Not determined.
Full Length
Pig
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers