missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Antibacterial peptide PMAP-36 Protein
A DNA sequence encoding the Sus scrofa (Pig) Antibacterial peptide PMAP-36, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001123437.1 |
---|---|
Gene ID (Entrez) | 100170124 |
Name | Antibacterial peptide PMAP-36 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017770
|
enQuireBio™
QP7219-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037770
|
enQuireBio™
QP7219-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027770
|
enQuireBio™
QP7219-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047770
|
enQuireBio™
QP7219-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057770
|
enQuireBio™
QP7219-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067770
|
enQuireBio™
QP7219-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001123437.1 | |
Antibacterial peptide PMAP-36 Protein | |
Not determined. | |
Full Length | |
Pig | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
100170124 | |
Research Use Only | |
PMAP36 | |
Recombinant Protein | |
E. coli | |
VGRFRRLRKK TRKRLKKIGK VLKWIPPIVG SIPLGCG | |
Greater than 90% as determined by SDS-PAGE. |