missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TIMP-1 / TIMP1 Protein
A DNA sequence encoding the Mus musculus (Mouse) TIMP-1 / TIMP1, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001037849.1 |
---|---|
Gene ID (Entrez) | 21857 |
Name | TIMP-1 / TIMP1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16095900
|
enQuireBio™
QP6795-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16015910
|
enQuireBio™
QP6795-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16005910
|
enQuireBio™
QP6795-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025910
|
enQuireBio™
QP6795-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035910
|
enQuireBio™
QP6795-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045910
|
enQuireBio™
QP6795-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001037849.1 | |
TIMP-1 / TIMP1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
21857 | |
Research Use Only | |
Timp1 | |
Recombinant Protein | |
E. coli | |
CSCAPPHPQT AFCNSDLVIR AKFMGSPEIN ETTLYQRYKI KMTKMLKGFK AVGNAADIRY AYTPVMESLC GYAHKSQNRS EEFLITGRLR NGNLHISACS FLVPWRTLSP AQQRAFSKTY SAGCGVCTVF PCLSIPCKLE SDTHCLWTDQ VLVGSEDYQS RHFACLPRNP GLCTWRSLGA R | |
Greater than 90% as determined by SDS-PAGE. |