missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TIMP-1 / TIMP1 Protein
A DNA sequence encoding the Mus musculus (Mouse) TIMP-1 / TIMP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6795-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001037849.1 | |
TIMP-1 / TIMP1 Protein | |
Research Use Only | |
Timp1 | |
Recombinant Protein | |
E. coli | |
CSCAPPHPQT AFCNSDLVIR AKFMGSPEIN ETTLYQRYKI KMTKMLKGFK AVGNAADIRY AYTPVMESLC GYAHKSQNRS EEFLITGRLR NGNLHISACS FLVPWRTLSP AQQRAFSKTY SAGCGVCTVF PCLSIPCKLE SDTHCLWTDQ VLVGSEDYQS RHFACLPRNP GLCTWRSLGA R | |
Greater than 90% as determined by SDS-PAGE. |
21857 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |