missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse SIRP beta 1a Fc Chimera Protein
Description
When Recombinant Mouse SIRP beta 1a Fc Chimera (Catalog # 10334-SB) is immobilized at 1 μg/mL (100 μL/well), Recombinant Human SP-D (Catalog # 1920-SP ) binds with an ED50 of 30-240 ng/mL.
Specifications
Specifications
| Accession Number | Q6F5F2 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 320832 |
| Molecular Weight (g/mol) | Observed: 83-92 kDa, under reducing conditions; Theroretical: 64 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse SIRP beta 1 A protein Mouse SIRP beta 1 A (Val28-Gly360) Accession # Q6F5F2(N-terminus) IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?