missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Serum Albumin / HSA / ALB Protein

A DNA sequence encoding the Mus musculus (Mouse) Serum Albumin / HSA / ALB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8965-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047321

  • 2325.00 EUR / 500µg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_033784.2
Serum Albumin / HSA / ALB Protein
500 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
11657
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ALB
Recombinant Protein
Yeast
EAHKSEIAHR YNDLGEQHFK GLVLIAFSQY LQKCSYDEHA KLVQEVTDFA KTCVADESAA NCDKSLHTLF GDKLCAIPNL RENYGELADC CTKQEPERNE CFLQHKDDNP SLPPFERPEA EAMCTSFKEN PTTFMGHYLH EVARRHPYFY APELLYYAEQ YNEILTQCCA EADKESCLTP KLDGVKEKAL VSSVRQRMKC SSMQKFGERA FKAWAVARLS QTFPNADFAE ITKLATDLTK VNKECCHGDL LECADDRAEL AKYMCENQAT ISSKLQTCCD KPLLKKAHCL SEVEHDTMPA DLPAIAADFV EDQEVCKNYA EAKDVFLGTF LYEYSRRHPD YSVSLLLRLA KKYEATLEKC CAEANPPACY GTVLAEFQPL VEEPKNLVKT NCDLYEKLGE YGFQNAILVR YTQKAPQVST PTLVEAARNL GRVGTKCCTL PEDQRLPCVE DYLSAILNRV CLLHEKTPVS EHVTKCCSGS LVERRPCFSA LTVDETYVPK EFKAETFTFH SDICTLPEKE KQIKKQTALA ELVKHKPKAT AEQLKTVMDD FAQFLDTCCK AADKDTCFST EGPNLVTRCK DALA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers