missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serum Albumin / HSA / ALB Protein
A DNA sequence encoding the Mus musculus (Mouse) Serum Albumin / HSA / ALB, was expressed in the hosts and tags indicated.
287.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_033784.2 |
---|---|
Gene ID (Entrez) | 11657 |
Name | Serum Albumin / HSA / ALB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16007321
|
enQuireBio™
QP8965-YE-100UG |
100 μg |
810.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047321
|
enQuireBio™
QP8965-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017321
|
enQuireBio™
QP8965-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037321
|
enQuireBio™
QP8965-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057321
|
enQuireBio™
QP8965-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027321
|
enQuireBio™
QP8965-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_033784.2 | |
Serum Albumin / HSA / ALB Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
11657 | |
Research Use Only | |
ALB | |
Recombinant Protein | |
Yeast | |
EAHKSEIAHR YNDLGEQHFK GLVLIAFSQY LQKCSYDEHA KLVQEVTDFA KTCVADESAA NCDKSLHTLF GDKLCAIPNL RENYGELADC CTKQEPERNE CFLQHKDDNP SLPPFERPEA EAMCTSFKEN PTTFMGHYLH EVARRHPYFY APELLYYAEQ YNEILTQCCA EADKESCLTP KLDGVKEKAL VSSVRQRMKC SSMQKFGERA FKAWAVARLS QTFPNADFAE ITKLATDLTK VNKECCHGDL LECADDRAEL AKYMCENQAT ISSKLQTCCD KPLLKKAHCL SEVEHDTMPA DLPAIAADFV EDQEVCKNYA EAKDVFLGTF LYEYSRRHPD YSVSLLLRLA KKYEATLEKC CAEANPPACY GTVLAEFQPL VEEPKNLVKT NCDLYEKLGE YGFQNAILVR YTQKAPQVST PTLVEAARNL GRVGTKCCTL PEDQRLPCVE DYLSAILNRV CLLHEKTPVS EHVTKCCSGS LVERRPCFSA LTVDETYVPK EFKAETFTFH SDICTLPEKE KQIKKQTALA ELVKHKPKAT AEQLKTVMDD FAQFLDTCCK AADKDTCFST EGPNLVTRCK DALA | |
Greater than 90% as determined by SDS-PAGE. |