missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NKX2.2 Protein
A DNA sequence encoding the Mus musculus (Mouse) NKX2.2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6423-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035049.1 | |
NKX2.2 Protein | |
Research Use Only | |
Nkx2-2 | |
Recombinant Protein | |
E. coli | |
MSLTNTKTGF SVKDILDLPD TNDEDGSVAE GPEEESEGPE PAKRAGPLGQ GALDAVQSLP LKSPFYDSSD NPYTRWLAST EGLQYSLHGL AASAPPQDSS SKSPEPSADE SPDNDKETQG GGGDAGKKRK RRVLFSKAQT YELERRFRQQ RYLSAPEREH LASLIRLTPT QVKIWFQNHR YKMKRARAEK GMEVTPLPSP RRVAVPVLVR DGKPCHALKA QDLAAATFQA GIPFSAYSAQ SLQHMQYNAQ YSSASTPQYP TAHPLVQAQQ WTW | |
Greater than 90% as determined by SDS-PAGE. |
18088 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |