missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NKX2.2 Protein
A DNA sequence encoding the Mus musculus (Mouse) NKX2.2, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_035049.1 |
---|---|
Gene ID (Entrez) | 18088 |
Name | NKX2.2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033210
|
enQuireBio™
QP6423-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053210
|
enQuireBio™
QP6423-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043210
|
enQuireBio™
QP6423-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063210
|
enQuireBio™
QP6423-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073210
|
enQuireBio™
QP6423-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083210
|
enQuireBio™
QP6423-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_035049.1 | |
NKX2.2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
18088 | |
Research Use Only | |
Nkx2-2 | |
Recombinant Protein | |
E. coli | |
MSLTNTKTGF SVKDILDLPD TNDEDGSVAE GPEEESEGPE PAKRAGPLGQ GALDAVQSLP LKSPFYDSSD NPYTRWLAST EGLQYSLHGL AASAPPQDSS SKSPEPSADE SPDNDKETQG GGGDAGKKRK RRVLFSKAQT YELERRFRQQ RYLSAPEREH LASLIRLTPT QVKIWFQNHR YKMKRARAEK GMEVTPLPSP RRVAVPVLVR DGKPCHALKA QDLAAATFQA GIPFSAYSAQ SLQHMQYNAQ YSSASTPQYP TAHPLVQAQQ WTW | |
Greater than 90% as determined by SDS-PAGE. |