missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MAPT / Tau Protein
A DNA sequence encoding the Mus musculus (Mouse) MAPT / Tau, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9264-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001033698.1 | |
MAPT / Tau Protein | |
Research Use Only | |
Mapt | |
Recombinant Protein | |
Yeast | |
ADPRQEFDTM EDHAGDYTLL QDQEGDMDHG LKESPPQPPA DDGAEEPGSE TSDAKSTPTA EDVTAPLVDE RAPDKQAAAQ PHTEIPEGIT AEEAGIGDTP NQEDQAAGHV TQGRREGQAP DLGTSDWTRQ QVSSMSGAPL LPQGLREATC QPSGTRPEDI EKSHPASELL RRGPPQKEGW GQDRLGSEEE VDEDLTVDES SQDSPPSQAS LTPGRAAPQA GSGSVCGETA SVPGLPTEGS VPLPADFFSK VSAETQASQP EGPGTGPMEE GHEAAPEFTF HVEIKASTPK EQDLEGATVV GVPGEEQKAQ TQGPSVGKGT KEASLQEPPG KQPAAGLPGR PVSRVPQLKA RVASKDRTGN DEKKAKTSTP SCAKAPSHRP CLSPTRPTLG SSDPLIKPSS PAVSPEPATS PKHVSSVTPR NGSPGTKQMK LKGADGKTGA KIATPRGAAS PAQKGTSNAT RIPAKTTPSP KTPPGSGEPP KSGERSGYSS PGSPGTPGSR SRTPSLPTPP TREPKKVAVV RTPPKSPSAS KSRLQTAPVP MPDLKNVRSK IGSTENLKHQ PGGGKVQIIN KKLDLSNVQS KCGSKDNIKH VPGGGSVQIV YKPVDLSKVT SKCGSLGNIH HKPGGGQVEV KSEKLDFKDR VQSKIGSLDN ITHVPGGGNK KIETHKLTFR ENAKAKTDHG AEIVYKSPVV SGDTSPRHLS NVSSTGSIDM VDSPQLATLA DEVSASLAKQ GL | |
Greater than 90% as determined by SDS-PAGE. |
17762 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |