missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MAPT / Tau Protein
A DNA sequence encoding the Mus musculus (Mouse) MAPT / Tau, was expressed in the hosts and tags indicated.
287.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_001033698.1 |
---|---|
Gene ID (Entrez) | 17762 |
Name | MAPT / Tau Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037951
|
enQuireBio™
QP9264-YE-100UG |
100 μg |
810.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077951
|
enQuireBio™
QP9264-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047951
|
enQuireBio™
QP9264-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067951
|
enQuireBio™
QP9264-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087951
|
enQuireBio™
QP9264-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057951
|
enQuireBio™
QP9264-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001033698.1 | |
MAPT / Tau Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
17762 | |
Research Use Only | |
Mapt | |
Recombinant Protein | |
Yeast | |
ADPRQEFDTM EDHAGDYTLL QDQEGDMDHG LKESPPQPPA DDGAEEPGSE TSDAKSTPTA EDVTAPLVDE RAPDKQAAAQ PHTEIPEGIT AEEAGIGDTP NQEDQAAGHV TQGRREGQAP DLGTSDWTRQ QVSSMSGAPL LPQGLREATC QPSGTRPEDI EKSHPASELL RRGPPQKEGW GQDRLGSEEE VDEDLTVDES SQDSPPSQAS LTPGRAAPQA GSGSVCGETA SVPGLPTEGS VPLPADFFSK VSAETQASQP EGPGTGPMEE GHEAAPEFTF HVEIKASTPK EQDLEGATVV GVPGEEQKAQ TQGPSVGKGT KEASLQEPPG KQPAAGLPGR PVSRVPQLKA RVASKDRTGN DEKKAKTSTP SCAKAPSHRP CLSPTRPTLG SSDPLIKPSS PAVSPEPATS PKHVSSVTPR NGSPGTKQMK LKGADGKTGA KIATPRGAAS PAQKGTSNAT RIPAKTTPSP KTPPGSGEPP KSGERSGYSS PGSPGTPGSR SRTPSLPTPP TREPKKVAVV RTPPKSPSAS KSRLQTAPVP MPDLKNVRSK IGSTENLKHQ PGGGKVQIIN KKLDLSNVQS KCGSKDNIKH VPGGGSVQIV YKPVDLSKVT SKCGSLGNIH HKPGGGQVEV KSEKLDFKDR VQSKIGSLDN ITHVPGGGNK KIETHKLTFR ENAKAKTDHG AEIVYKSPVV SGDTSPRHLS NVSSTGSIDM VDSPQLATLA DEVSASLAKQ GL | |
Greater than 90% as determined by SDS-PAGE. |