missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Interferon regulatory factor 5 Protein
A DNA sequence encoding the Mus musculus (Mouse) Interferon regulatory factor 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6234-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001239311.1 | |
Interferon regulatory factor 5 Protein | |
Research Use Only | |
Irf5 | |
Recombinant Protein | |
E. coli | |
MNHSAPGIPP PPRRVRLKPW LVAQVNSCQY PGLQWVNGEK KLFYIPWRHA TRHGPSQDGD NTIFKAWAKE TGKYTEGVDE ADPAKWKANL RCALNKSRDF QLFYDGPRDM PPQPYKIYEV CSNGPAPTES QPTDDYVLGE EEEEEEEELQ RMLPGLSITE PALPGPPNAP YSLPKEDTKW PPALQPPVGL GPPVPDPNLL APPSGNPAGF RQLLPEVLEP GPLASSQPPT EPLLPDLLIS PHMLPLTDLE IKFQYRGRAP RTLTISNPQG CRLFYSQLEA TQEQVELFGP VTLEQVRFPS PEDIPSDKQR FYTNQLLDVL DRGLILQLQG QDLYAIRLCQ CKVFWSGPCA LAHGSCPNPI QREVKTKLFS LEQFLNELIL FQKGQTNTPP PFEIFFCFGE EWPDVKPREK KLITVQVVPV AARLLLEMFS GELSWSADSI RLQISNPDLK DHMVEQFKEL HHLWQSQQQL QPMVQAPPVA GLDASQGPWP MHPVGMQ | |
Greater than 90% as determined by SDS-PAGE. |
27056 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |