missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Interferon regulatory factor 5 Protein
A DNA sequence encoding the Mus musculus (Mouse) Interferon regulatory factor 5, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001239311.1 |
---|---|
Gene ID (Entrez) | 27056 |
Name | Interferon regulatory factor 5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031800
|
enQuireBio™
QP6234-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051800
|
enQuireBio™
QP6234-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041800
|
enQuireBio™
QP6234-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061800
|
enQuireBio™
QP6234-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071800
|
enQuireBio™
QP6234-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081800
|
enQuireBio™
QP6234-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001239311.1 | |
Interferon regulatory factor 5 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
27056 | |
Research Use Only | |
Irf5 | |
Recombinant Protein | |
E. coli | |
MNHSAPGIPP PPRRVRLKPW LVAQVNSCQY PGLQWVNGEK KLFYIPWRHA TRHGPSQDGD NTIFKAWAKE TGKYTEGVDE ADPAKWKANL RCALNKSRDF QLFYDGPRDM PPQPYKIYEV CSNGPAPTES QPTDDYVLGE EEEEEEEELQ RMLPGLSITE PALPGPPNAP YSLPKEDTKW PPALQPPVGL GPPVPDPNLL APPSGNPAGF RQLLPEVLEP GPLASSQPPT EPLLPDLLIS PHMLPLTDLE IKFQYRGRAP RTLTISNPQG CRLFYSQLEA TQEQVELFGP VTLEQVRFPS PEDIPSDKQR FYTNQLLDVL DRGLILQLQG QDLYAIRLCQ CKVFWSGPCA LAHGSCPNPI QREVKTKLFS LEQFLNELIL FQKGQTNTPP PFEIFFCFGE EWPDVKPREK KLITVQVVPV AARLLLEMFS GELSWSADSI RLQISNPDLK DHMVEQFKEL HHLWQSQQQL QPMVQAPPVA GLDASQGPWP MHPVGMQ | |
Greater than 90% as determined by SDS-PAGE. |