missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Interferon regulatory factor 5 Protein

A DNA sequence encoding the Mus musculus (Mouse) Interferon regulatory factor 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6234-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041800

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001239311.1
Interferon regulatory factor 5 Protein
Research Use Only
Irf5
Recombinant Protein
E. coli
MNHSAPGIPP PPRRVRLKPW LVAQVNSCQY PGLQWVNGEK KLFYIPWRHA TRHGPSQDGD NTIFKAWAKE TGKYTEGVDE ADPAKWKANL RCALNKSRDF QLFYDGPRDM PPQPYKIYEV CSNGPAPTES QPTDDYVLGE EEEEEEEELQ RMLPGLSITE PALPGPPNAP YSLPKEDTKW PPALQPPVGL GPPVPDPNLL APPSGNPAGF RQLLPEVLEP GPLASSQPPT EPLLPDLLIS PHMLPLTDLE IKFQYRGRAP RTLTISNPQG CRLFYSQLEA TQEQVELFGP VTLEQVRFPS PEDIPSDKQR FYTNQLLDVL DRGLILQLQG QDLYAIRLCQ CKVFWSGPCA LAHGSCPNPI QREVKTKLFS LEQFLNELIL FQKGQTNTPP PFEIFFCFGE EWPDVKPREK KLITVQVVPV AARLLLEMFS GELSWSADSI RLQISNPDLK DHMVEQFKEL HHLWQSQQQL QPMVQAPPVA GLDASQGPWP MHPVGMQ
Greater than 90% as determined by SDS-PAGE.
27056
10 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers