missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Interferon regulatory factor 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Interferon regulatory factor 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9220-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_058545.1 | |
Interferon regulatory factor 3 Protein | |
Research Use Only | |
Irf3 | |
Recombinant Protein | |
Yeast | |
METPKPRILP WLVSQLDLGQ LEGVAWLDES RTRFRIPWKH GLRQDAQMAD FGIFQAWAEA SGAYTPGKDK PDVSTWKRNF RSALNRKEVL RLAADNSKDP YDPHKVYEFV TPGARDFVHL GASPDTNGKS SLPHSQENLP KLFDGLILGP LKDEGSSDLA IVSDPSQQLP SPNVNNFLNP APQENPLKQL LAEEQWEFEV TAFYRGRQVF QQTLFCPGGL RLVGSTADMT LPWQPVTLPD PEGFLTDKLV KEYVGQVLKG LGNGLALWQA GQCLWAQRLG HSHAFWALGE ELLPDSGRGP DGEVHKDKDG AVFDLRPFVA DLIAFMEGSG HSPRYTLWFC MGEMWPQDQP WVKRLVMVKV VPTCLKELLE MAREGGASSL KTVDLHISNS QPISLTSDQY KAYLQDLVED MDFQATGNI | |
Greater than 90% as determined by SDS-PAGE. |
54131 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |