missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Interferon regulatory factor 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Interferon regulatory factor 3, was expressed in the hosts and tags indicated.
245.00 EUR - 2645.00 EUR
Specifications
Accession Number | NP_058545.1 |
---|---|
Gene ID (Entrez) | 54131 |
Name | Interferon regulatory factor 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057861
|
enQuireBio™
QP9220-YE-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007871
|
enQuireBio™
QP9220-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067861
|
enQuireBio™
QP9220-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087861
|
enQuireBio™
QP9220-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097861
|
enQuireBio™
QP9220-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077861
|
enQuireBio™
QP9220-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_058545.1 | |
Interferon regulatory factor 3 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
54131 | |
Research Use Only | |
Irf3 | |
Recombinant Protein | |
Yeast | |
METPKPRILP WLVSQLDLGQ LEGVAWLDES RTRFRIPWKH GLRQDAQMAD FGIFQAWAEA SGAYTPGKDK PDVSTWKRNF RSALNRKEVL RLAADNSKDP YDPHKVYEFV TPGARDFVHL GASPDTNGKS SLPHSQENLP KLFDGLILGP LKDEGSSDLA IVSDPSQQLP SPNVNNFLNP APQENPLKQL LAEEQWEFEV TAFYRGRQVF QQTLFCPGGL RLVGSTADMT LPWQPVTLPD PEGFLTDKLV KEYVGQVLKG LGNGLALWQA GQCLWAQRLG HSHAFWALGE ELLPDSGRGP DGEVHKDKDG AVFDLRPFVA DLIAFMEGSG HSPRYTLWFC MGEMWPQDQP WVKRLVMVKV VPTCLKELLE MAREGGASSL KTVDLHISNS QPISLTSDQY KAYLQDLVED MDFQATGNI | |
Greater than 90% as determined by SDS-PAGE. |