Learn More
R&D Systems™ Recombinant Mouse Integrin alpha V beta 6 Protein
Click to view available options
:
50ug; Carrier Free
Description
The Recombinant Mouse Integrin alpha V beta 6 Protein is derived from CHO. The Recombinant Mouse Integrin alpha V beta 6 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 115 kDa ( alphaV subunit) & 82.6 kDa (beta6 subunit) |
| Name | Integrin alpha V beta 6 |
| Quantity | 50μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha V beta 6 protein, (N-terminus) Mouse Integrin alpha V (Phe31-Val988) Accession &Num; P43406 HP GGGSGGGSGGGS Acidic Tail HHHHHH Mouse Integrin beta 6 (Gly22-Asn706) Accession &Num; Q9Z0T9 GGGSGGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Non-covalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |