missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IFN-alpha 11 Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
10 μg
50 μg
missing translation for 'unitSize'
10µg
50µg
Description
Vogel, S.N. et al. (1982) Infect. Immunol. 38:681. The ED50 for this effect is <20 pg/mL.
Specifications
Specifications
| Accession Number | Q61716.2 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 15964 |
| Molecular Weight (g/mol) | Observed: 18-24 kDa, under reducing conditions; Theroretical: 19 kDa |
| Quantity | 10 μg |
| Source | Human embryonic kidney cell, HEK293-derived mouse IFN-alpha 11 protein Cys24-Ala190 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 100 μg/mL in PBS. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction