missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse H2-K1 Protein
A DNA sequence encoding the Mus musculus (Mouse) H2-K1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7268-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
H2-K1 Protein | |
Research Use Only | |
H2-K1 | |
Recombinant Protein | |
E. coli | |
GPHSLRYFVT AVSRPGLGEP RFIAVGYVDD TQFVRFDSDA DNPRFEPRAP WMEQEGPEYW EEQTQRAKSD EQWFRVSLRT AQRYYNQSKG GSHTFQRMFG CDVGSDWRLL RGYQQFAYDG RDYIALNEDL KTWTAADTAA LITRRKWEQA GDAEYYRAYL EGECVEWLRR YLELGNETLL RTDSPKAHVT YHPRSQVDVT LRCWALGFYP ADITLTWQLN GEDLTQDMEL VETRPAGDGT FQKWAAVVVP LGKEQNYTCH VHHKGLPEPL TLRWKLPPST VSNT | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |