missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse H2-K1 Protein
A DNA sequence encoding the Mus musculus (Mouse) H2-K1, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Name | H2-K1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | H2-K1 |
Protein Length | Extracellular Domain |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057940
|
enQuireBio™
QP7268-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077940
|
enQuireBio™
QP7268-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067940
|
enQuireBio™
QP7268-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087940
|
enQuireBio™
QP7268-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097940
|
enQuireBio™
QP7268-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007950
|
enQuireBio™
QP7268-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
H2-K1 Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
H2-K1 | |
Recombinant Protein | |
E. coli | |
GPHSLRYFVT AVSRPGLGEP RFIAVGYVDD TQFVRFDSDA DNPRFEPRAP WMEQEGPEYW EEQTQRAKSD EQWFRVSLRT AQRYYNQSKG GSHTFQRMFG CDVGSDWRLL RGYQQFAYDG RDYIALNEDL KTWTAADTAA LITRRKWEQA GDAEYYRAYL EGECVEWLRR YLELGNETLL RTDSPKAHVT YHPRSQVDVT LRCWALGFYP ADITLTWQLN GEDLTQDMEL VETRPAGDGT FQKWAAVVVP LGKEQNYTCH VHHKGLPEPL TLRWKLPPST VSNT | |
Greater than 90% as determined by SDS-PAGE. |