missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Group XIIA secretory phospholipase A2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Group XIIA secretory phospholipase A2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8063-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042451

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_075685.2
Group XIIA secretory phospholipase A2 Protein
Research Use Only
PLA2G12A
Recombinant Protein
E. coli
QEQDQTTDWR ATLKTIRNGI HKIDTYLNAA LDLLGGEDGL CQYKCSDGSK PVPRYGYKPS PPNGCGSPLF GVHLNIGIPS LTKCCNQHDR CYETCGKSKN DCDEEFQYCL SKICRDVQKT LGLSQNVQAC ETTVELLFDS VIHLGCKPYL DSQRAACWCR YEEKTDL
Greater than 90% as determined by SDS-PAGE.
66350
500 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers