missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Group XIIA secretory phospholipase A2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Group XIIA secretory phospholipase A2, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_075685.2 |
---|---|
Gene ID (Entrez) | 66350 |
Name | Group XIIA secretory phospholipase A2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002451
|
enQuireBio™
QP8063-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022451
|
enQuireBio™
QP8063-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16012451
|
enQuireBio™
QP8063-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032451
|
enQuireBio™
QP8063-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042451
|
enQuireBio™
QP8063-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052451
|
enQuireBio™
QP8063-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_075685.2 | |
Group XIIA secretory phospholipase A2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
66350 | |
Research Use Only | |
PLA2G12A | |
Recombinant Protein | |
E. coli | |
QEQDQTTDWR ATLKTIRNGI HKIDTYLNAA LDLLGGEDGL CQYKCSDGSK PVPRYGYKPS PPNGCGSPLF GVHLNIGIPS LTKCCNQHDR CYETCGKSKN DCDEEFQYCL SKICRDVQKT LGLSQNVQAC ETTVELLFDS VIHLGCKPYL DSQRAACWCR YEEKTDL | |
Greater than 90% as determined by SDS-PAGE. |