missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Group XIIA secretory phospholipase A2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Group XIIA secretory phospholipase A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8063-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_075685.2 | |
Group XIIA secretory phospholipase A2 Protein | |
Research Use Only | |
PLA2G12A | |
Recombinant Protein | |
E. coli | |
QEQDQTTDWR ATLKTIRNGI HKIDTYLNAA LDLLGGEDGL CQYKCSDGSK PVPRYGYKPS PPNGCGSPLF GVHLNIGIPS LTKCCNQHDR CYETCGKSKN DCDEEFQYCL SKICRDVQKT LGLSQNVQAC ETTVELLFDS VIHLGCKPYL DSQRAACWCR YEEKTDL | |
Greater than 90% as determined by SDS-PAGE. |
66350 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |