missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Collagenase 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Collagenase 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8902-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032633.1 | |
Collagenase 3 Protein | |
Research Use Only | |
MMP13 | |
Recombinant Protein | |
E. coli | |
YNVFPRTLKW SQTNLTYRIV NYTPDMSHSE VEKAFRKAFK VWSDVTPLNF TRIYDGTADI MISFGTKEHG DFYPFDGPSG LLAHAFPPGP NYGGDAHFDD DETWTSSSKG YNLFIVAAHE LGHSLGLDHS KDPGALMFPI YTYTGKSHFM LPDDDVQGIQ FLYGPGDEDP NPKHPKTPEK CDPALSLDAI TSLRGETMIF KDRFFWRLHP QQVEAELFLT KSFWPELPNH VDAAYEHPSR DLMFIFRGRK FWALNGYDIL EGYPRKISDL GFPKEVKRLS AAVHFENTGK TLFFSENHVW SYDDVNQTMD KDYPRLIEEE FPGIGNKVDA VYEKNGYIYF FNGPIQFEYS IWSNRIVRVM PTNSILWC | |
Greater than 90% as determined by SDS-PAGE. |
17386 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |