missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Collagenase 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Collagenase 3, was expressed in the hosts and tags indicated.
245.00 EUR - 2645.00 EUR
Specifications
Accession Number | NP_032633.1 |
---|---|
Gene ID (Entrez) | 17386 |
Name | Collagenase 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16067091
|
enQuireBio™
QP8902-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007101
|
enQuireBio™
QP8902-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057091
|
enQuireBio™
QP8902-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087091
|
enQuireBio™
QP8902-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097091
|
enQuireBio™
QP8902-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077091
|
enQuireBio™
QP8902-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_032633.1 | |
Collagenase 3 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
17386 | |
Research Use Only | |
MMP13 | |
Recombinant Protein | |
E. coli | |
YNVFPRTLKW SQTNLTYRIV NYTPDMSHSE VEKAFRKAFK VWSDVTPLNF TRIYDGTADI MISFGTKEHG DFYPFDGPSG LLAHAFPPGP NYGGDAHFDD DETWTSSSKG YNLFIVAAHE LGHSLGLDHS KDPGALMFPI YTYTGKSHFM LPDDDVQGIQ FLYGPGDEDP NPKHPKTPEK CDPALSLDAI TSLRGETMIF KDRFFWRLHP QQVEAELFLT KSFWPELPNH VDAAYEHPSR DLMFIFRGRK FWALNGYDIL EGYPRKISDL GFPKEVKRLS AAVHFENTGK TLFFSENHVW SYDDVNQTMD KDYPRLIEEE FPGIGNKVDA VYEKNGYIYF FNGPIQFEYS IWSNRIVRVM PTNSILWC | |
Greater than 90% as determined by SDS-PAGE. |