Learn More
R&D Systems™ Recombinant Mouse Carboxyl Ester Lipase/CEL Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
10 μg
missing translation for 'unitSize'
10µg
Description
The Recombinant Mouse Carboxyl Ester Lipase/CEL Protein is derived from NS0. The Recombinant Mouse Carboxyl Ester Lipase/CEL Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 12613 |
| Molecular Weight (g/mol) | 58 kDa |
| Name | Carboxyl Ester Lipase/CEL |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Carboxyl Ester Lipase/CEL protein Ala21-Leu534,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Structural Form | Monomer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |