missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse C-C motif chemokine 9 Protein

A DNA sequence encoding the Mus musculus (Mouse) C-C motif chemokine 9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8647-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16085741

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_035468.1
C-C motif chemokine 9 Protein
Research Use Only
Ccl9
Recombinant Protein
E. coli
QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQN
Greater than 90% as determined by SDS-PAGE.
20308
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers