missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C-C motif chemokine 9 Protein
A DNA sequence encoding the Mus musculus (Mouse) C-C motif chemokine 9, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_035468.1 |
---|---|
Gene ID (Entrez) | 20308 |
Name | C-C motif chemokine 9 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075741
|
enQuireBio™
QP8647-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16095741
|
enQuireBio™
QP8647-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16085741
|
enQuireBio™
QP8647-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005751
|
enQuireBio™
QP8647-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015751
|
enQuireBio™
QP8647-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025751
|
enQuireBio™
QP8647-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_035468.1 | |
C-C motif chemokine 9 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20308 | |
Research Use Only | |
Ccl9 | |
Recombinant Protein | |
E. coli | |
QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQN | |
Greater than 90% as determined by SDS-PAGE. |