missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C-C motif chemokine 9 Protein
A DNA sequence encoding the Mus musculus (Mouse) C-C motif chemokine 9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8647-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035468.1 | |
C-C motif chemokine 9 Protein | |
Research Use Only | |
Ccl9 | |
Recombinant Protein | |
E. coli | |
QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQN | |
Greater than 90% as determined by SDS-PAGE. |
20308 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |