Learn More
enQuireBio™ Recombinant M. tuberculosis ESAT-6-like protein EsxH Protein
Product Code. 16088570
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16088570
Brand: enQuireBio™ QP7384ec100ug
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) ESAT-6-like protein EsxH, was expressed in the hosts and tags indicated.
Specifications
Accession Number WP_003401514.1
Name M. tuberculosis ESAT-6-like protein EsxH Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol esxH
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity M. tuberculosis
Species E. coli
Protein Tag His-SUMO
Sequence SQIMYNYPAM LGHAGDMAGY AGTLQSLGAE IAVEQAALQS AWQGDTGITY QAWQAQWNQA MEDLVRAYHA MSSTHEANTM AMMARDTAEA AKWGG
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.