missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza B (strain B / Singapore / 222 / 1979) NucleoProtein
A DNA sequence encoding the Influenza B virus (strain B/Singapore/222/1979) Influenza B (strain B/Singapore/222/1979) Nucleoprotein, was expressed in the hosts and tags indicated.
309.00 EUR - 2645.00 EUR
Specifications
Name | Influenza B (strain B / Singapore / 222 / 1979) NucleoProtein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | NP |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088010
|
enQuireBio™
QP7285-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028020
|
enQuireBio™
QP7285-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078010
|
enQuireBio™
QP7285-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008020
|
enQuireBio™
QP7285-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018020
|
enQuireBio™
QP7285-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098010
|
enQuireBio™
QP7285-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Influenza B (strain B / Singapore / 222 / 1979) NucleoProtein | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MSNMDIDGIN TGTIDKTPEE IISGTSGATR PIIRPATLAP PSNKRTRNPS PERATTSSEA DVGRKTQKKQ TPTEIKKSVY NMVVKLGEFY NQMMVKAGLN DDMERNLIQN AHAVERILLA ATDDKKTEFQ KKKNARDVKE GKEEIDHNKT GGTFYKMVRD DKTIYFSPIR ITFLKEEVKT MYKTTMGSDG FSGLNHIMIG HSQMNDVCFQ RSKALKRVGL DPSLISTFAG STLPRRSGAT GVAIKGGGTL VAEAIRFIGR AMADRGLLRD IKAKTAYEKI LLNLKNKCSA PQQKALVDQV IGSRNPGIAD IEDLTLLARS MVVVRPSVAS KVVLPISIYA KIPQLGFNVE EYSMVGYEAM ALYNMATPVS ILRMGDDAKD KSQLFFMSCF GAAYEDLRVL SALTGTEFKP RSALKCKGFH VPAKEQVEGM GAALMSIKLQ FWAPMTRSGG NEVGGDGGSG QISCSPVFAV ERPIALSKQA VRRMLSMNIE GRDADVKGNL LKMMNDSMAK KTNGNAFIGK KMFQISDKNK TNPVEIPIKQ TIPNFFFGRD TAEDYDDLDY | |
Greater than 90% as determined by SDS-PAGE. |