missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza B (strain B / Singapore / 222 / 1979) NucleoProtein
A DNA sequence encoding the Influenza B virus (strain B/Singapore/222/1979) Influenza B (strain B/Singapore/222/1979) Nucleoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7285-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
Influenza B (strain B / Singapore / 222 / 1979) NucleoProtein | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MSNMDIDGIN TGTIDKTPEE IISGTSGATR PIIRPATLAP PSNKRTRNPS PERATTSSEA DVGRKTQKKQ TPTEIKKSVY NMVVKLGEFY NQMMVKAGLN DDMERNLIQN AHAVERILLA ATDDKKTEFQ KKKNARDVKE GKEEIDHNKT GGTFYKMVRD DKTIYFSPIR ITFLKEEVKT MYKTTMGSDG FSGLNHIMIG HSQMNDVCFQ RSKALKRVGL DPSLISTFAG STLPRRSGAT GVAIKGGGTL VAEAIRFIGR AMADRGLLRD IKAKTAYEKI LLNLKNKCSA PQQKALVDQV IGSRNPGIAD IEDLTLLARS MVVVRPSVAS KVVLPISIYA KIPQLGFNVE EYSMVGYEAM ALYNMATPVS ILRMGDDAKD KSQLFFMSCF GAAYEDLRVL SALTGTEFKP RSALKCKGFH VPAKEQVEGM GAALMSIKLQ FWAPMTRSGG NEVGGDGGSG QISCSPVFAV ERPIALSKQA VRRMLSMNIE GRDADVKGNL LKMMNDSMAK KTNGNAFIGK KMFQISDKNK TNPVEIPIKQ TIPNFFFGRD TAEDYDDLDY | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |