missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Influenza A Virus H1N1 Neuraminidase Protein
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
Description
Measured by its ability to cleave a fluorogenic substrate, 2’-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid. The specific activity is >2,500 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | AAF77036 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. |
| Molecular Weight (g/mol) | M.W. (Observed): 57 kDa, reducing conditions; M.W. (theoretical): 48 kDa |
| Name | Viral Neuraminidase |
| Quantity | 5 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived influenza a virus h1n1 Viral Neuraminidase protein Ser37-Lys469, with an N-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction